ENA321763598 Enamine

2-methyl-3,4-dihydro-2H-1-benzothiopyran-4-one

  • MDL number MFCD12131644
  • Cas Number : 826-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18-19
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75377
Availability details for
Quantity
  • Availability
  • Add to Cart