ENA321763602 Enamine

1-(1-methylpiperidin-4-yl)pyrrolidin-3-amine

  • MDL number MFCD12167337
  • Cas Number : 1096892-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75381
Availability details for
Quantity
  • Availability
  • Add to Cart