ENA321763607 Enamine

7-fluoro-4-methyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD09736748
  • Cas Number : 749240-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75386
Availability details for
Quantity
  • Availability
  • Add to Cart