ENA321763609 Enamine

3-(pyridin-4-yloxy)aniline

  • MDL number MFCD06825512
  • Cas Number : 102877-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75389

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75389
Availability details for
Quantity
  • Availability
  • Add to Cart