ENA321763613 Enamine

2-(thiophen-3-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD11208448
  • Cas Number : 1094385-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75401
Availability details for
Quantity
  • Availability
  • Add to Cart