ENA321763616 Enamine

2-(benzylamino)acetamide

  • MDL number MFCD06858463
  • Cas Number : 39796-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75404

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75404
Availability details for
Quantity
  • Availability
  • Add to Cart