ENA321763620 Enamine

methyl 2-(phenylamino)butanoate

  • MDL number MFCD12775604
  • Cas Number : 77165-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75409

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75409
Availability details for
Quantity
  • Availability
  • Add to Cart