ENA321763628 Enamine

1-{2-[(4-bromobenzene)sulfonyl]ethyl}piperidin-3-amine

  • MDL number MFCD13336272
  • Cas Number : 1303755-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75421

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75421
Availability details for
Quantity
  • Availability
  • Add to Cart