ENA321763634 Enamine

2-chloro-N,N-dimethylpyridine-4-carboxamide

  • MDL number MFCD03411707
  • Cas Number : 209262-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75427
Availability details for
Quantity
  • Availability
  • Add to Cart