ENA321763637 Enamine

N-(3-nitro-1,4-dihydropyridin-4-ylidene)-1-(oxolan-2-yl)methanamine

  • Cas Number : 1040064-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74516
Availability details for
Quantity
  • Availability
  • Add to Cart