ENA321763638 Enamine

4-N-[1-(oxolan-2-yl)ethyl]pyridine-3,4-diamine

  • MDL number MFCD12195332
  • Cas Number : 1153405-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75431

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75431
Availability details for
Quantity
  • Availability
  • Add to Cart