ENA321763639 Enamine

2-benzylbutanoic acid

  • MDL number MFCD01722150
  • Cas Number : 5669-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75432
Availability details for
Quantity
  • Availability
  • Add to Cart