ENA321763640 Enamine

2-bromo-1-(4,5-dihydrofuran-3-yl)-2,2-difluoroethan-1-one

  • MDL number MFCD11099669
  • Cas Number : 1087792-68-4
  • Molecular Weight : 227.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75433
Availability details for
Quantity
  • Availability
  • Add to Cart