ENA321763641 Enamine

3-methyl-4-phenoxybenzaldehyde

  • MDL number MFCD16717339
  • Cas Number : 191284-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75434
Availability details for
Quantity
  • Availability
  • Add to Cart