ENA321763643 Enamine

1-[1-(propan-2-yl)-1H-pyrazol-4-yl]ethan-1-amine

  • MDL number MFCD08700769
  • Cas Number : 1007459-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75436
Availability details for
Quantity
  • Availability
  • Add to Cart