ENA321763646 Enamine

3-(4-acetylpiperazin-1-yl)propanoic acid

  • MDL number MFCD04116700
  • Cas Number : 705941-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74519
Availability details for
Quantity
  • Availability
  • Add to Cart