ENA321763647 Enamine

N-(cyclopropylmethyl)-2-(2-formylphenoxy)acetamide

  • MDL number MFCD11149392
  • Cas Number : 1020948-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75441
Availability details for
Quantity
  • Availability
  • Add to Cart