ENA321763649 Enamine

2-(cyclopentylamino)pyridine-3-carboxylic acid

  • MDL number MFCD03114311
  • Cas Number : 460363-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75443
Availability details for
Quantity
  • Availability
  • Add to Cart