ENA321763652 Enamine

5-(morpholin-4-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00233546
  • Cas Number : 37454-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 329-330
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75447
Availability details for
Quantity
  • Availability
  • Add to Cart