ENA321763660 Enamine

1-ethylimidazolidin-2-one

  • MDL number MFCD11226854
  • Cas Number : 872-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75465

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75465
Availability details for
Quantity
  • Availability
  • Add to Cart