ENA321763672 Enamine

1,3-diethyl 2-propanoylpropanedioate

  • MDL number MFCD00234681
  • Cas Number : 21633-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75481

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-75481
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart