ENA321763676 Enamine

ethyl 3-(4-fluoro-3-methylphenyl)-3-oxopropanoate

  • MDL number MFCD07783536
  • Cas Number : 72835-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75486

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75486
Availability details for
Quantity
  • Availability
  • Add to Cart