ENA321763678 Enamine

2-chloro-1-(4,5-dihydrofuran-3-yl)-2,2-difluoroethan-1-one

  • MDL number MFCD11099670
  • Cas Number : 1087784-77-7
  • Molecular Weight : 182.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75494
Availability details for
Quantity
  • Availability
  • Add to Cart