ENA321763680 Enamine

tert-butyl N-(2-{[(4-methylbenzene)sulfonyl]oxy}ethyl)carbamate

  • MDL number MFCD07776965
  • Cas Number : 158690-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75497

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-75497
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart