ENA321763682 Enamine

2-methyl-5-sulfamoylbenzamide

  • MDL number MFCD11646531
  • Cas Number : 76503-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75499
Availability details for
Quantity
  • Availability
  • Add to Cart