ENA321763687 Enamine

2-[4-(propane-2-sulfonyl)phenyl]acetic acid

  • MDL number MFCD11986718
  • Cas Number : 1105193-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75506

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75506
Availability details for
Quantity
  • Availability
  • Add to Cart