ENA321763688 Enamine

2-{[(2,5-dimethoxyphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD11175324
  • Cas Number : 40171-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91...93
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74336

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74336
Availability details for
Quantity
  • Availability
  • Add to Cart