ENA321763689 Enamine

1-[(piperidin-4-yl)methyl]-2,3-dihydro-1H-indole

  • MDL number MFCD06801219
  • Cas Number : 883549-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-74539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74539
Availability details for
Quantity
  • Availability
  • Add to Cart