ENA321763690 Enamine

N-(4-chlorophenyl)-2-{[(pyridin-3-yl)methyl]amino}acetamide

  • MDL number MFCD12549929
  • Cas Number : 1040053-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75509
Availability details for
Quantity
  • Availability
  • Add to Cart