ENA321763696 Enamine

cyclopropyl(2-methylphenyl)methanamine

  • MDL number MFCD05191952
  • Cas Number : 535925-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75516

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75516
Availability details for
Quantity
  • Availability
  • Add to Cart