ENA321763710 Enamine

3-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)-3-oxopropanenitrile

  • MDL number MFCD12142668
  • Cas Number : 1154379-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75546
Availability details for
Quantity
  • Availability
  • Add to Cart