ENA321763717 Enamine

2-cyano-N-cyclopropyl-N-(2,2,2-trifluoroethyl)acetamide

  • MDL number MFCD16059327
  • Cas Number : 1248497-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-75554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75554
Availability details for
Quantity
  • Availability
  • Add to Cart