ENA321763718 Enamine

2-(5-methyl-1,3-thiazol-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD18071422
  • Cas Number : 1311317-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75555

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75555
Availability details for
Quantity
  • Availability
  • Add to Cart