ENA321763725 Enamine

1-(3,4-dichlorophenyl)-2-(2,3-dihydro-1H-inden-5-yloxy)ethan-1-one

  • MDL number MFCD13686565
  • Cas Number : 1154284-20-4
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75563
Availability details for
Quantity
  • Availability
  • Add to Cart