ENA321763732 Enamine

1-(1-benzofuran-2-yl)-2,2-dimethylpropan-1-one

  • MDL number MFCD11174126
  • Cas Number : 75566-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75572
Availability details for
Quantity
  • Availability
  • Add to Cart