ENA321763736 Enamine

N-(4-hydroxycyclohexyl)-1H-1,3-benzodiazole-5-carboxamide

  • MDL number MFCD12471138
  • Cas Number : 1156718-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74660

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74660
Availability details for
Quantity
  • Availability
  • Add to Cart