ENA321763738 Enamine

1-[(4-bromo-2-fluorophenyl)methyl]-4-methyl-1,4-diazepane

  • MDL number MFCD13227270
  • Cas Number : 1291624-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75578
Availability details for
Quantity
  • Availability
  • Add to Cart