ENA321763751 Enamine

1-(3-amino-4-chlorophenyl)imidazolidin-2-one

  • MDL number MFCD11167806
  • Cas Number : 1038282-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75616

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75616
Availability details for
Quantity
  • Availability
  • Add to Cart