ENA321763754 Enamine

2-amino-1-(4-hydroxypiperidin-1-yl)-2-methylpropan-1-one hydrochloride

  • MDL number MFCD13562108
  • Cas Number : 1220033-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252...254
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75620

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75620
Availability details for
Quantity
  • Availability
  • Add to Cart