ENA321763760 Enamine

1-(2-hydroxypropyl)piperidin-4-ol

  • MDL number MFCD12802195
  • Cas Number : 1183328-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75627

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75627
Availability details for
Quantity
  • Availability
  • Add to Cart