ENA321763775 Enamine

2-(3-chlorophenoxy)-2-methylpropanoic acid

  • MDL number MFCD01695660
  • Cas Number : 17413-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75658
Availability details for
Quantity
  • Availability
  • Add to Cart