ENA321763778 Enamine

1-isocyanato-2-methoxyethane

  • MDL number MFCD09673759
  • Cas Number : 42170-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75661

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-75661
pictograms GHS02,GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart