ENA321763784 Enamine

2-methyl-3-(methylsulfanyl)propan-1-amine

  • MDL number MFCD12823762
  • Cas Number : 1183068-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75667
Availability details for
Quantity
  • Availability
  • Add to Cart