ENA321763798 Enamine

1-{2-[(2-aminoethyl)sulfanyl]ethoxy}-3-bromobenzene

  • MDL number MFCD11577801
  • Cas Number : 1082443-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74683

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74683
Availability details for
Quantity
  • Availability
  • Add to Cart