ENA321763799 Enamine

2-(thiophene-2-amido)thiophene-3-carboxylic acid

  • MDL number MFCD09047860
  • Cas Number : 926235-66-7
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75682
Availability details for
Quantity
  • Availability
  • Add to Cart