ENA321763805 Enamine

2-(3-bromopropoxy)-1,3-dimethylbenzene

  • MDL number MFCD00596680
  • Cas Number : 3245-54-3
  • Molecular Weight : 243.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-75689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75689
Availability details for
Quantity
  • Availability
  • Add to Cart