ENA321763807 Enamine

1-[(pyridin-4-yl)methyl]piperidine-4-carboxylic acid dihydrochloride

  • MDL number MFCD06801262
  • Cas Number : 1172453-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75692
Availability details for
Quantity
  • Availability
  • Add to Cart