ENA321763821 Enamine

3-[(1,2,3,4-tetrahydroisoquinoline-2-carbonyl)amino]propanoic acid

  • MDL number MFCD10011924
  • Cas Number : 1017667-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-74348
Availability details for
Quantity
  • Availability
  • Add to Cart