ENA321763824 Enamine

1-methyl-4-phenylpiperidin-4-ol

  • MDL number MFCD00464382
  • Cas Number : 4972-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75728

Safety Information

msds https://www.enaminestore.com/catalog/EN300-75728
Availability details for
Quantity
  • Availability
  • Add to Cart