ENA321763826 Enamine

1-(4-amino-2-methylphenyl)pyrrolidine-2,5-dione

  • MDL number MFCD09736330
  • Cas Number : 91091-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-75730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75730
Availability details for
Quantity
  • Availability
  • Add to Cart